Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora A/B/C Antibody (4A7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB360491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360491 Supplier Novus Biologicals Supplier No. NBP250046AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora A/B/C Monoclonal antibody specifically detects Aurora A/B/C in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Aurora A/B/C
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 4A7
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AIE2, AIK, AIK2, AIK3, AIM1, AIM-1, ARK1, ARK2, ARK-2, ARK3, ARK-3, AURA, AurB, AurC, aurkb-sv1, aurkb-sv2, aurora 2, aurora 3, aurora- and Ipl1-like midbody-associated protein 1, aurora kinase A, aurora kinase B, aurora kinase B-Sv1, aurora kinase B-Sv2, aurora kinase C, aurora/IPL1/EG2 protein 2, aurora/IPL1-like kinase, aurora/IPL1-related kinase 1, aurora/IPL1-related kinase 2, aurora/IPL1-related kinase 3, aurora-1, aurora-B, aurora-C, aurora-related kinase 2, aurora-related kinase 3, breast tumor-amplified kinase, BTAK, epididymis secretory protein Li 90, HEL-S-90, IPL1, PPP1R47, PPP1R48, protein phosphatase 1, regulatory subunit 47, protein phosphatase 1, regulatory subunit 48, serine/threonine kinase 12, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine protein kinase 15, serine/threonine-protein kinase 12, serine/threonine-protein kinase 13, serine/threonine-protein kinase 5, serine/threonine-protein kinase 6, serine/threonine-protein kinase aurora-A, serine/thr
Host Species Mouse
Immunogen Full length recombinant human AURKA protein expressed from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.