Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aurora A/B Monoclonal Antibody (5A12)
Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | Aurora A/B |
---|---|
Clone | 5A12 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547340
|
Invitrogen™
MA547340 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547439
|
Invitrogen™
MA547439 |
100 μL |
Each for $526.00
|
|
|||||
Description
Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.Specifications
Aurora A/B | |
1 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
O14965, O55099, O70126, P59241, P97477, Q96GD4 | |
AURKA, AURKB | |
Primary | |
Protein A/G |
5A12 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse, Rat, Canine, Horse | |
AURKA | |
100050161, 100073050, 114592, 20877, 20878, 261730, 479492, 485940, 6790, 9212 | |
Full length recombinant human Aurora A protein expressed in and purified from E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title