Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Antibody (3F11), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB360530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360530 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360530 Supplier Novus Biologicals Supplier No. NBP250039DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora B Monoclonal antibody specifically detects Aurora B in Human, Mouse, Rat, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay

Specifications

Antigen Aurora B
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Proximity Ligation Assay
Classification Monoclonal
Clone 3F11
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
Formulation 50mM Sodium Borate
Gene Alias AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5
Host Species Mouse
Immunogen Full length recombinant human aurora B protein expressed in and purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9212
Target Species Human, Mouse, Rat, Bovine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.