Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Antibody (AURKB/3121R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB382069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382069 Supplier Novus Biologicals Supplier No. NBP308280JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Aurora B Monoclonal antibody specifically detects Aurora B in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Aurora B
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AURKB/3121R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5
Host Species Rabbit
Immunogen Recombinant fragment (around aa 89-251) of human Aurora B protein (exact sequence is proprietary) (Uniprot: Q96GD4)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.