Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Rabbit anti-Human, Mouse, Rat, Clone: 1N9V5, Novus Biologicals™
SDP

Catalog No. NB166620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166620 20 μg
NB166619 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166620 Supplier Novus Biologicals Supplier No. NBP31576220UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Aurora B Monoclonal antibody specifically detects Aurora B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Aurora B
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1N9V5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Aurora B (NP_004208.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9212
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.