Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora C Antibody (OTI2B7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271303A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271303A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271303A48 Supplier Novus Biologicals Supplier No. NBP271303AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora C Monoclonal specifically detects Aurora C in Human samples. It is validated for Western Blot.

Specifications

Antigen Aurora C
Applications Western Blot
Classification Monoclonal
Clone OTI2B7
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3
Gene Symbols AURKC
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Aurora C expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6795.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.