Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora C Antibody (OTI2B7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288966 Supplier Novus Biologicals Supplier No. NBP271303APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora C Monoclonal antibody specifically detects Aurora C in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Aurora C
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B7
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human AURKC expression vector
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6795
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.