Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahlstrom-Munksjö AutoCollect™

Catalog No. 12007318 Shop All Ahlstrom Products
Change view
Click to view available options
Format:
Cassette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
12-007-318 Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-007-318 Supplier Ahlstrom-Munksjö Supplier No. 8.560.0003.BN
Add to Cart
Add to Cart

High quality specimen collection cards designed for laboratory automated equipment

  • For microvolume sampling, transport and storage of dried blood spots (DBS) at ambient temperature
  • Cassette format of card with high stiffness provides suitable fit with main laboratory automated equipment available on market
    • Includes: Camag DBS-MS 500, Hamilton EasyPunch and Spark Holland DBS Autosampler
  • Allowing reliable DBS card processing from detection of dried blood spot to prep of samples ready for LC/MS analysis

Advantages of Automation:

  • Traceability of operations
  • Volume control
  • Accuracy of timing
  • Analysis of results instead of results acquisition
  • Can be used routinely in high-throughput environments
  • Demonstrated to work efficiently in DBS extraction or direct elution
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For microvolume sampling, transport and storage of dried blood spots in ambient temps
For Use With (Equipment) Laboratory automated equipment
Format Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.