Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Autoimmune Regulator/AIRE Antibody (609930) [FITC], Novus Biologicals™
SDP

Catalog No. NB376928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376928 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376928 Supplier Novus Biologicals Supplier No. FAB6184F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Autoimmune Regulator/AIRE Monoclonal antibody specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Autoimmune Regulator/AIRE
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 609930
Conjugate FITC
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias APECED protein, APECEDAPSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1
Host Species Rat
Immunogen E. coli-derived recombinant mouse AIRE, Ser476-Ser552 (predicted), Accession # Q9Z0E3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 326
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.