Learn More
Thermo Scientific Chemicals Autophagy Inhibitor, 3-MA
Description
A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation

Specifications
Specifications
Quantity | 25 mg |
Description | White |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation |
Formula Weight | 149.15g/mol |
Product Type | Signal Transduction Reagent |
Form | Solid |
MDL Number | MFCD00010531 |
Beilstein | 146087 |
InChI Key | FSASIHFSFGAIJM-UHFFFAOYSA-N |
Show More |
Safety and Handling
EINECSNumber | 225-908-6 |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.