Learn More
Alfa Aesar™ Autophagy Inhibitor, 3-MA
Manufacturer:  Alfa Aesar™ J64813MB
 View more versions of this product
Catalog No. AAJ64813MB
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
Specifications
Specifications
25mg
2° to 8°C
A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
C6H7N5
3-Methyladenine; 6-Amino-3-methylpurine
White
Solid
5142-23-4
149.15g/mol
Signal Transduction Reagent