Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Auxin response factor Mouse anti-Plant, Clone: 14D2, Novus Biologicals™
SDP

Catalog No. NB036700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036700 Supplier Novus Biologicals Supplier No. NBP289117

Mouse Monoclonal Antibody

Auxin response factor Monoclonal specifically detects Auxin response factor in Plant samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Auxin response factor
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14D2
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Host Species Mouse
Immunogen Recombinant protein within tea Auxin response factor aa 401-586/ 820. (SwissProt: J7KE95 Tea)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4442
Target Species Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.