Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avi Epitope Tag Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB293780 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293780 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293780 Supplier Novus Biologicals Supplier No. NBP326066F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avi Epitope Tag Polyclonal antibody specifically detects Avi Epitope Tag in Epitope Tag samples. It is validated for Western Blot

Specifications

Antigen Avi Epitope Tag
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the Avi Epitope Tag sequence (GLNDIFEAQKIEWHE)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.