Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Flu M1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001933SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001933SS 0.025 mg
NB1001933 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001933SS Supplier Novus Biologicals Supplier No. NB1001933SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Flu M1 Polyclonal specifically detects Avian Flu M1 in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Avian Flu M1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Alias Avian Influenza Matrix Protein 1
Host Species Rabbit
Immunogen Amino acids 9-28 of avian flu matrix protein 1 were used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen has 100% identity to the H5N1 strain in chicken, duck and swine.
Target Species Virus, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.