Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Flu M1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10056610

 View more versions of this product

Catalog No. NB10056610


Only null left
Add to Cart

Description

Description

Avian Flu M1 Polyclonal antibody specifically detects Avian Flu M1 in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Avian Flu M1
Polyclonal
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Rabbit
Protein G purified
RUO
The amino acid sequence used as immunogen is 100% homologous to duck H5N1 strain and 92% homologous to chicken H5N1 strain.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 0.5-2 ug/ml
Avian Influenza Matrix Protein 1
Amino acids 212-225 of avian flu matrix protein 1 were used as the immunogen.
0.1 mg
Primary
Virus, Chicken
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only