Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Flu M1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Avian Flu M1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056610T
![]() |
Novus Biologicals
NB10056610SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056610
![]() |
Novus Biologicals
NB10056610 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Avian Flu Matrix Protein 1 Polyclonal specifically detects Avian Flu Matrix Protein 1 in Avian, Chicken samples. It is validated for Western Blot.Specifications
Avian Flu M1 | |
Western Blot 0.5-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
Amino acids 212-225 of avian flu matrix protein 1 were used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Virus, Chicken | |
Avian Influenza Matrix Protein 1 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous to duck H5N1 strain and 92% homologous to chicken H5N1 strain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title