Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H5N1 Hemagglutinin Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB220294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB220294 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB220294 Supplier Novus Biologicals Supplier No. NBP241065AF532
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Avian Influenza A H5N1 Hemagglutinin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Harvey rat sarcoma viral oncogene homolog
Host Species Goat
Immunogen Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the center of the Hemagglutinin protein Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 250 - 300 of Avian Influenza Hemagglutinin 3. Amino Acid Squence: APEYAYKIVKKGDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.