Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H5N1 M2 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB223011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB223011 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB223011 Supplier Novus Biologicals Supplier No. NBP241074DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza A H5N1 M2 Polyclonal antibody specifically detects Avian Influenza A H5N1 M2 in Virus samples. It is validated for ELISA

Specifications

Antigen Avian Influenza A H5N1 M2
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from amino terminus of H5N1 M2. The immunogen is located within the first 50 amino acids of Avian Influenza A M2. Amino Acid Squence: LTEVETPTRNEWE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.