Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H5N1 Neuraminidase Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB220024 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB220024 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB220024 Supplier Novus Biologicals Supplier No. NBP241048AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISA

Specifications

Antigen Avian Influenza A H5N1 Neuraminidase
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 16 amino acids in the middle of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within amino acids 150 - 200 of Avian Influenza Neuraminidase. Amino Acid Squence: CPIGESPSPYNSRFES
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.