Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H5N1 Neuraminidase Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB222821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222821 Supplier Novus Biologicals Supplier No. NBP241049H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISA

Specifications

Antigen Avian Influenza A H5N1 Neuraminidase
Applications ELISA
Classification Polyclonal
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Neuraminidase. Amino Acid Squence: IRGRPKENTIWTSGS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.