Learn More
Description
Specifications
Specifications
| Antigen | Avian Influenza A H6N1 Nucleoprotein |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA 0.1-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Thompson et al (2006)) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | NP, Nucleoprotein |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 428-441 (AFTGNTEGRTSDMR) of avian flu nucleoprotein were used as the immunogen, GenBank no ADC34563.1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
