Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H6N1 Nucleoprotein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056572T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056572T 0.025 mg
NB10056572 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056572T Supplier Novus Biologicals Supplier No. NB10056572SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Avian Influenza A H6N1 Nucleoprotein Polyclonal specifically detects Avian Influenza A H6N1 Nucleoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Avian Influenza A H6N1 Nucleoprotein
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution ELISA 0.1-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Thompson et al (2006))
Formulation PBS with 0.05% Sodium Azide
Gene Alias NP, Nucleoprotein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 428-441 (AFTGNTEGRTSDMR) of avian flu nucleoprotein were used as the immunogen, GenBank no ADC34563.1.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen has 100% identity to the H5N1 virus in chicken, duck and swine.
Target Species Virus, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.