Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Avian Influenza A H7N9 Hemagglutinin Antibody Pair [HRP]
SDP

Catalog No. p-7183081 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin.

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin.
Target Species Virus
For Use With (Application) Sandwich ELISA
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 110 ng of recombinant Avian Influenza A H7N9 Hemagglutinin.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-Avian Influenza A H7N9 Hemagglutinin monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL rabbit anti-Avian Influenza A H7N9 Hemagglutinin polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Avian Influenza A H7N9 Hemagglutinin
    Technique 1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.