Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H7N9 Neuraminidase Antibody - (A/Shanghai/02/2013) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396426 0.025 mg
NBP241278 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396426 Supplier Novus Biologicals Supplier No. NBP2412780.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza A H7N9 Neuraminidase Polyclonal specifically detects Avian Influenza A H7N9 Neuraminidase in Virus samples. It is validated for ELISA.

Specifications

Antigen Avian Influenza A H7N9 Neuraminidase
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias EC=3.2.1.18, Neuraminidase
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 11 amino acids near the amino terminus of the H7N9 [Influenza A virus (A/Shanghai/02/2013(H7N9))] Neuraminidase protein. The immunogen is located within amino acids 50 - 100 of Avian Influenza A H7N9 Neuraminidase. Amino Acid Squence: QMEERTSRNFN
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity virus
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.