Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H7N9 Neuraminidase Antibody - (A/Shanghai/02/2013) - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Avian Influenza A H7N9 Neuraminidase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Avian Influenza A H7N9 Neuraminidase Polyclonal specifically detects Avian Influenza A H7N9 Neuraminidase in Virus samples. It is validated for ELISA.Specifications
Avian Influenza A H7N9 Neuraminidase | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide corresponding to 11 amino acids near the amino terminus of the H7N9 [Influenza A virus (A/Shanghai/02/2013(H7N9))] Neuraminidase protein. The immunogen is located within amino acids 50 - 100 of Avian Influenza A H7N9 Neuraminidase. Amino Acid Squence: QMEERTSRNFN | |
Primary | |
virus |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Virus | |
EC=3.2.1.18, Neuraminidase | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title