Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza NS1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB222564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222564 Supplier Novus Biologicals Supplier No. NBP241067JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza NS1 Polyclonal antibody specifically detects Avian Influenza NS1 in Virus samples. It is validated for ELISA

Specifications

Antigen Avian Influenza NS1
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 13.6kDa protein, 14.5kDa protein, Control protein orf2, Control protein orf3, Control protein orf4, E4, E4 13.5kDa protein, E4 13.6kDa protein, E4 14.4kDa protein, E4 14.5kDa protein, E4 control protein 34 kDa, E4 control protein orf2, E4 control protein orf3, E4 control protein orf4, HuAdVD
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the carboxy terminus of the Avian Influenza Nonstructural Protein 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Nonstructural Protein 1. Amino Acid Squence: LPPNQKRKMARTIE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.