Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPI1 Antibody (OTI1G1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272121A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272121A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272121A94 Supplier Novus Biologicals Supplier No. NBP272121AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AVPI1 Monoclonal specifically detects AVPI1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen AVPI1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G1
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias arginine vasopressin-induced 1, arginine vasopressin-induced protein 1, AVP-induced protein 1, PP5395, RP11-548K23.7, vasopressin-induced transcript, VIP32, VIT32
Gene Symbols AVPI1
Host Species Mouse
Immunogen Full length recombinant protein of human AVPI1 (NP_068378) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 60370
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.