Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axin-1 Antibody (4E9F1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61695 Supplier Novus Biologicals Supplier No. NBP261695
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Axin-1 Monoclonal specifically detects Axin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Axin-1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4E9F1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250
Formulation PBS with 0.05% Sodium Azide
Gene Alias axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315
Gene Symbols AXIN1
Host Species Mouse
Immunogen Purified recombinant fragment of human AXIN1 (AA: 546-752) expressed in E. Coli.
Molecular Weight of Antigen 95.6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8312
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.