Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axin-1 Antibody (4E9F1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Axin-1 |
---|---|
Clone | 4E9F1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61695
![]() |
Novus Biologicals
NBP261695 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
Axin-1 Monoclonal specifically detects Axin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Axin-1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
AXIN1 | |
IgG2b | |
Protein G purified |
4E9F1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
Monoclonal | |
Purified | |
Apoptosis, Wnt Signaling Pathway | |
8312 | |
Purified recombinant fragment of human AXIN1 (AA: 546-752) expressed in E. Coli. | |
Primary | |
95.6 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title