Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axin-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358192 Supplier Novus Biologicals Supplier No. NBP176483MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Axin-1 Polyclonal antibody specifically detects Axin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Axin-1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human AXIN1. The immunogen is located within amino acids 620 - 670 of AXIN1. Amino Acid Squence: EKEISRHRRTGHGS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8312
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.