Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Axitinib

Catalog No. AC463800010
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AC463800010 1 g
AC463802500 250 mg
2 options

Catalog No. AC463800010

Supplier: Thermo Scientific Chemicals 463800010

Only null left
Add to Cart

CAS: 319460-85-0 | C22H18N4OS | 386.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319460-85-0
Molecular Formula C22H18N4OS
Molecular Weight (g/mol) 386.47
InChI Key RITAVMQDGBJQJZ-FMIVXFBMSA-N
IUPAC Name N-methyl-2-({3-[(1E)-2-(pyridin-2-yl)ethenyl]-1H-indazol-6-yl}sulfanyl)benzamide
SMILES CNC(=O)C1=CC=CC=C1SC1=CC=C2C(NN=C2\C=C\C2=CC=CC=N2)=C1

Specifications

Melting Point 226.0°C to 228.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol, dimethylsulfoxide
Formula Weight 386.47
Chemical Name or Material Axitinib
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.