Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axl Antibody (108724R) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB399609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399609 Supplier Novus Biologicals Supplier No. FAB154RL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF

Specifications

Antigen Axl
Applications Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
Classification Monoclonal
Clone 108724R
Conjugate DyLight 550
Dilution Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Axl, Met1-Pro440, Accession # AAA61243
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 558
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.