Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axl Antibody (175128), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310734 Supplier Novus Biologicals Supplier No. FAB8541PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Axl Monoclonal antibody specifically detects Axl in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen Axl
Applications Flow Cytometry
Classification Monoclonal
Clone 175128
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Axl, Ala19-Trp445, Accession # Q00993
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 558
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.