Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axl Antibody (MM0098-2N33) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2120440.025MG
Description
Axl Monoclonal specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface), CyTOF-ready.Specifications
Axl | |
Monoclonal | |
LYOPH | |
Flow Cytometry 1:50-100, Flow (Cell Surface), CyTOF-ready | |
AAA61243 | |
AXL | |
Human recombinant Axl EC domain | |
0.025 mg | |
Cell Cycle and Replication | |
558 | |
Human, Mouse | |
Purified |
Flow Cytometry, CyTOF | |
MM0098-2N33 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction