Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axl Antibody (MM0098-2N33) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | Axl |
|---|---|
| Clone | MM0098-2N33 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 1:50-100, Flow (Cell Surface), CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
Description
Axl Monoclonal specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface), CyTOF-ready.Specifications
| Axl | |
| LYOPH | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| AAA61243 | |
| 558 | |
| Human recombinant Axl EC domain | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0098-2N33 | |
| Flow Cytometry 1:50-100, Flow (Cell Surface), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| AXL | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title