Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axl Antibody (MM0098-2N33), PE, Novus Biologicals™
SDP

Catalog No. NBP212044PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212044PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212044PE Supplier Novus Biologicals Supplier No. NBP212044PE

Mouse Monoclonal Antibody

Axl Monoclonal specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen Axl
Applications Flow Cytometry
Classification Monoclonal
Clone MM0098-2N33
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. AAA61243
Gene Alias AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
Gene Symbols AXL
Host Species Mouse
Immunogen Human recombinant Axl EC domain
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 558
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.