Learn More
Invitrogen™ Axl Monoclonal Antibody (7E10)

Description
MA5-15504 targets AXL in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15504 immunogen is purified recombinant extracellular fragment of human AXL fused with hIgGFc tag expressed in HEK293 cell line. MA5-15504 detects AXL which has a predicted molecular weight of approximately 97.4kDa.
Specifications
Specifications
| Antigen | Axl |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 7E10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | AXL |
| Gene Accession No. | P30530 |
| Gene Alias | Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.