Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axotrophin Antibody, Novus Biologicals™
SDP

Catalog No. NBP15526220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15526220 20 μL
NBP155262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15526220 Supplier Novus Biologicals Supplier No. NBP15526220UL

Rabbit Polyclonal Antibody

Axotrophin Polyclonal specifically detects Axotrophin in Human samples. It is validated for Western Blot.

Specifications

Antigen Axotrophin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q9H992
Gene Alias AXO, AXOT, Axotrophin, E3 ubiquitin-protein ligase MARCH7, EC 6.3.2, EC 6.3.2.-, MARCH-VIIDKFZp586F1122, membrane-associated ring finger (C3HC4) 7, Membrane-associated RING finger protein 7, Membrane-associated RING-CH protein VII, RING finger protein 177, RNF177axotrophin
Gene Symbols 43166
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MARCH7(membrane-associated ring finger (C3HC4) 7) The peptide sequence was selected from the middle region of 40244. Peptide sequence SSMSSTFFSRRSSQDSLNTRSLNSENSYVSPRILTASQSRSNVPSASEVP.
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 64844
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.