Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Caps for PCR Tubes
GSA_VAGREENER_CHOICE

Catalog No. 14222265
Click to view available options
Color:
Assorted
Clear
MaxyClear
MaxyClear, w/writing surface
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps

8-Strip domed or flat strip caps for Axygen PCR tubes

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Color Clear
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Strip
Product Type 8-Strip Domed PCR Tube Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.