Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Gel Documentation System Accessory, Optional Filters

Catalog No. p-5270603
Click to view available options
Wavelength:
497nm
572nm
676nm
Color:
Blue
Green
Red

For use with Gel Documentation System (GDBL-1000)

Note: For research use only. Not intended for use in diagnostic or therapeutic procedures.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Gel Documentation System (GDBL-1000)
Color Blue
Wavelength 497nm
Product Type Filter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.