Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Axygen™ Microplate Sealing Film and Tapes
Shop All Axygen Products
                    Click to view available options
                
                
                    
                            
                                Material:
                            
                        
                        
                            
                                Aluminum
                            
                        
                            
                                Litto or Ethylene or Vinyl
                            
                        
                            
                                Polyester
                            
                        
                            
                                Polyolefin
                            
                        
                            
                                Rayon
                            
                        
                    
                            
                                Product Type:
                            
                        
                        
                            
                                AxySeal Sealing Film
                            
                        
                            
                                Breathable Sealing Film
                            
                        
                            
                                CyclerSeal Sealing Film
                            
                        
                            
                                CyclerSeal Sealing Film Roll
                            
                        
                            
                                Heat Seal Film
                            
                        
                            
                                Sealing Film
                            
                        
                            
                                Sealing Film Roll
                            
                        
                            
                                UltraClear Sealing Film
                            
                        
                    
                            
                                Dimensions (L x W):
                            
                        
                        
                            
                                115 x 500 m
                            
                        
                            
                                127.2 x 79.5 mm
                            
                        
                            
                                133.3 x 79.2 mm
                            
                        
                            
                                135.5 x 80 mm
                            
                        
                            
                                146 x 79.2 mm
                            
                        
                            
                                146 x 79.6 mm
                            
                        
                            
                                153 x 82.3 mm
                            
                        
                            
                                Not Available
                            
                        
                    
                            
                                Sterility:
                            
                        
                        
                            
                                Non-sterile
                            
                        
                            
                                Sterile
                            
                        
                    Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging
 
            Specifications
Specifications
| Material | Litto or Ethylene or Vinyl | 
| Sterility | Non-sterile | 
| Length (English) | 5 in. | 
| Length (Metric) | 127.2 mm | 
| No. per Case | 100/Cs. | 
| No. per Pack | 100/Pk. | 
| Dimensions (L x W) | 127.2 x 79.5 mm | 
| Width (English) | 3.12 in. | 
| Width (Metric) | 79.5 mm | 
| For Use With (Application) | Designed for applications ranging from PCR and real-time PCR to storage | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            