Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Microplate Sealing Film and Tapes
Shop All Axygen Products
Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
CyclerSeal Sealing Film Roll
Heat Seal Film
Sealing Film
Sealing Film Roll
UltraClear Sealing Film
Dimensions (L x W):
115 x 500 m
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
Not Available
Sterility:
Non-sterile
Sterile
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging
Specifications
Specifications
| Material | Polyester |
| Sterility | Non-sterile |
| Length (English) | 4.52 in. |
| Length (Metric) | 115 mm |
| No. per Case | 1/Cs. |
| No. per Pack | 1/Pk. |
| Dimensions (L x W) | 115 x 500 m |
| Width (English) | 196 in. |
| Width (Metric) | 500 m |
| For Use With (Application) | Designed for applications ranging from PCR and real-time PCR to storage |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction