Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Catalog No. p-4419052 Shop All Axygen Products
Click to view available options
Product Type:
Heat Sealing Film for Compound Storage
Heat Sealing Film for qPCR
Peelable Heat Sealing Film
Peelable Heat Sealing Film for qPCR
Pierceable Aluminum Heat Sealing Film
This item is not returnable. View return policy
This item is not returnable. View return policy
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Plate Sealing
For Use With (Equipment) PlateMax Plate Sealer
Material Polyester
No. per Case 500/Cs.
No. per Pack 100/Pk.
Product Type Heat Sealing Film for qPCR
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.