Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE

Catalog No. 14222965 Shop All Axygen Products
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
25 Tubes and Caps/Unit, 20 Units/Cs.
50 Tubes and Caps/Unit, 20 Units/Cs.
Bag

Graduated tubes with conical bottom

  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Closure Material HDPE
Autoclavable Not Autoclavable
Capacity (Metric) 50 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
No. per Pack 250/Pk.
Packaging 25 Tubes and Caps/Rack, 20 Racks/Cs.
Sterility Sterile
Graduated Yes
Closure Type Screw Cap
Bottom Shape Cylindrical
Max. RCF 12,000 x g
Product Type Centrifuge Tube
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.