Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Catalog No. 14222239 Shop All Axygen Products
Click to view available options
Color:
Blue
Clear
Sterility:
Non-sterile
Sterile
No. of Wells:
24
48
96
384
Volume (Metric) Well:
1.1 mL
1.8 mL
10,000 μL
1100 μL
125 μL
2000 μL
240 μL
5000 μL
550 μL

For sample collection, in-vitro growth chambers and long term storage.

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Array 8 x 12
    Autoclavable Autoclavable
    Color Blue
    Material Polypropylene
    Volume (Metric) Well 550 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Conical Bottom (V-well)
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.