Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AZ 10397767

Click to view available options
Quantity:
10 mg
50 mg
Description
AZ 10397767 is a potent CXCR2 antagonist (IC50 = 1 nM); attenuates oxaliplatin -induced NF-κB activation, increases oxaliplatin cytotoxicity, and potentiates oxaliplatin-induced apoptosis in AIPC cells. Reduces the numbers of neutrophils infiltrating into tumors in both in vitro and in vivo models and delayed tumor growth. Orally bioavailable.
Specifications
Specifications
CAS | 333742-63-5 |
Quantity | 10 mg |
Target | Chemokine Receptor |
Molecular Formula | C15H14ClFN4O2S2 |
Formula Weight | 400.88g/mol |
Product Type | Antagonist |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction