Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 11645373
GREENER_CHOICE

Catalog No. 331710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-171-0 10 mg
33-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-171-0 Supplier Tocris Bioscience™ Supplier No. 3317/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective human P2X7 antagonist

Potent and selective human P2X7 antagonist (KB values are 5 - 7 and > 10,000 nM at hP2X7 and rP2X7 respectively) that is completely without effect at all other P2X subtypes. Inhibits BzATP-mediated calcium influx and inhibits ATP-mediated IL-1β release in vitro (KB values are 15 and 92 nM respectively).

Chemical Identifiers

CAS 227088-94-0
Molecular Formula C24H21N3O5S

Specifications

Quantity 10 mg
Formula Weight 463.51
Purity >98%
Molecular Weight (g/mol) 463.51
Chemical Name or Material AZ 11645373
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.