Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AZ 13705339

Click to view available options
Quantity:
10 mg
50 mg
Description
AZ 13705339 is a highly potent PAK1 and PAK2 inhibitor (IC50 values are 0.33 and 6 nM for PAK1 and PAK2, respectively). Displays >7500-fold selectivity for PAK1 over PAK4 (IC50 = 2.6 μM).
Specifications
Specifications
| Chemical Name or Material | 2-[[[2-[[3-(Ethylsulfonyl)-4-(4-methyl-1-piperazinyl)phenyl]amino]-5-fluoro-4-pyrimidinyl][5-(hydroxymethyl)-2-methylphenyl]amino]methyl]benzonitrile |
| CAS | 2016806-57-6 |
| Quantity | 10 mg |
| Molecular Formula | C33H36FN7O3S |
| Formula Weight | 629.75 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction