Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AZ 20

Click to view available options
Quantity:
10 mg
50 mg
Description
AZ 20 is a potent and selective ATR kinase inhibitor (IC50 = 5 nM). Exhibits 7.6-fold selectivity over mTOR and selectivity over a panel of 442 kinases, including ATM kinase, PI3-K isoforms, and DNA-PK. Inhibits cell growth in cell lines with high baseline levels of replication stress. Displays antitumor effects in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | AZ 20 |
| Purity | >98% |
| Molecular Weight (g/mol) | 412.51 |
| Inhibitors | AZ 20 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction