Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ PFKFB3 26
GREENER_CHOICE

Catalog No. 567510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-751-0 10 mg
56-755-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-751-0 Supplier Tocris Bioscience™ Supplier No. 5675/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PFKFB3 inhibitor

Potent and selective PFKFB3 inhibitor (IC50 = 0.023 μM). Selective for PFKFB3 over PFKFB2 and PFKFB1 isoforms (IC50 values are 0.382 μM and 2.06 μM, respectively). Reduces levels of fructose-1,6-bisphosphate in A549 cells (IC50 = 0.343 μM). Exhibits no significant inhibition of other kinases.

Chemical Identifiers

CAS 1704740-52-2
Molecular Formula C24H26N4O2

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 402.49
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 402.498
Chemical Name or Material (2S)-N-[4-[[3-Cyano-1-(2-methylpropyl)-1H-indol-5-yl]oxy]phenyl]-2-pyrrolidinecarboxamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.